Fetal Hemoglobin Kit, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
Modified Kleihauer-Betke procedure
M-Clarity™ Qualitätsstufe = MQ500
285D-1KT
285D-1KTEA
453
EUR
285D-1KT
Fetal Hemoglobin Kit, Sigma-Aldrich®
Fetal Hemoglobin Kit
Amyloid protein is detected in tissue with Congo red, a metachromatic stain. Staining is intensified by pretreatment of tissue with alkaline sodium chloride.
- Suitable for haematology
- Suitable for histology
- Sufficient for 200 manual assays
Kit component also available separately
Citrate Phosphate Buffer Concentrate 2 x 100 mL (2851)
Acid hematoxylin solution 2 x 100 mL (2852)
Eosin B solution, 0.1% aqueous solution 2 x 100 mL (2853)
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...