Toluol ≥99,90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zur Analyse

Lieferant: VWR Chemicals
Gefahr

Synonyme: Anisen, Methylbenzol, Methylbenzen, Toluen, Retinnaphtha, Phenylmethan

28676.297EA 56.9 EUR
28676.297 28676.322 28676.366 28676.468 28676.550 28676.950
Toluol ≥99,90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zur Analyse
Toluol
Formel: C₆H₅CH₃
Molecular Weight: 92.14 g/mol
Siedepunkt: 110.6 °C (1013 hPa)
Schmelzpunkt: –95 °C
Dichte: 0.867 g/cm³ (20 °C)
Flammpunkt: 4
Lagertemperatur: Raumtemperatur
MDL: MFCD00008512
CAS-Nummer: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 01-2119471310-51
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Clear colourless liquid
Gehalt (ber. auf wasserfreie Substanz) Min. 99,90 %
Carbonisable substances Passes test
IR-Spektrum Passes test
Substances coloured by H2SO4 Passes test ACS/ISO
Acidity or alkalinity Max. 0.0001 meq/g
Boiling point 109 - 112 °C
Colouration Max. 10 APHA
Density (20/4) 0.865 - 0.868
Density (20/20) 0.865 - 0.870
Brechungsindex (20°C) 1.496 - 1.498
Benzene Max. 50 ppm
Evaporation residue Max. 10 ppm
Thiophene Max. 1 ppm
Total S (as SO4) Max. 10 ppm
Water Max. 0.03 %
Al (Aluminium) Max. 0.05 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.2 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.01 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.05 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.01 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR