D(+)-Trehalose Dihydrat ≥98% für die Biochemie

Lieferant: VWR Chemicals

Synonyme: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate

28719.290EA 583 EUR
28719.290
D(+)-Trehalose Dihydrat ≥98% für die Biochemie
D(+)-Trehalose Dihydrat
Formel: C₁₂H₂₂O₁₁·2H₂O
Molecular Weight: 378.33 g/mol
Schmelzpunkt: 97…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00071594
CAS-Nummer: 6138-23-4
EINECS: 202-739-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af741ee934bfe7c82395d31c21e2c19f0738d0f0&vid=59183c854dcfa367e15d46861ef4b9a7028540a9&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Min. 98 %
Ignition residue (SO4) Max. 0.1 %
Cl (Chloride) Max. 50 ppm
SO4 (Sulphate) Max. 50 ppm
As (Arsenic) Max. 0.1 ppm
Cd (Cadmium) Max. 5 ppm
Co (Cobalt) Max. 5 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 5 ppm
Ni (Nickel) Max. 5 ppm
Pb (Lead) Max. 5 ppm
Zn (Zinc) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR