Schwefelsäure 95,0-98,0%, AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr


Veränderungsmanagement-Kategorie = N

2876-21EA 0 EUR
2876-21 2876-28
Schwefelsäure 95,0-98,0%, AR® ACS, Macron Fine Chemicals™
Schwefelsäure
Formel: H₂SO₄
Siedepunkt: ∼335 °C (1013 hPa)
Schmelzpunkt: 3 °C
Dichte: 1,84 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064589
CAS-Nummer: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use Passes Test
Appearance Passes Test
ACS - Assay (H₂SO₄) 95.0 - 98.0 %
ACS - Color (APHA) ≤10
ACS - Residue after Ignition ≤5 ppm
ACS - Chloride (Cl) ≤0.2 ppm
ACS - Nitrate (NO₃) ≤0.5 ppm
ACS - Ammonium (NH₄) ≤2 ppm
ACS - Substances Reducing Permanganate (as SO₂) ≤2 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Mercury (Hg) ≤5.0 ppb
Trace Impurities - Arsenic (As) ≤0.010 ppm
Trace Impurities - Boron (B) ≤0.010 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR