2',5'-Didesoxyadenosin, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-methyltetrahydrofuran-3-ol

288104-1
288104-1EA 101 EUR
288104-1
2',5'-Didesoxyadenosin, Millipore®
2',5'-Didesoxyadenosin
Formel: C₁₀H₁₃N₅O₂
Molecular Weight: 235,2425 g/mol
MDL: MFCD00210904
CAS-Nummer: 6698-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR