L(-)-Tryptophan ≥99% für die Biochemie
Lieferant: VWR Chemicals
Synonyme:
L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
28821.131EA
179
EUR
28821.131
L(-)-Tryptophan ≥99% für die Biochemie
L(-)-Tryptophan
Formel:
C₁₁H₁₂N₂O₂ Molecular Weight: 204,23 g/mol Schmelzpunkt: 290 °C Dichte: 1,34 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064340 CAS-Nummer: 73-22-3 EINECS: 200-795-6 Merck Index: 12,09929 |
Spezifikation Testergebnisse
Assay (HClO4) (on dried substance) | ≥ 99 % |
Identification (IR) | Passes test |
Identification (specific rotation) | Passes test |
Appearance of solution (10 g/l; HCl 1 M) | Clear and colourless |
Specif. opt. rotation (10 g/l; on dried) | -33.0 - -30.0 ° |
Ninhydrin-positive substances (any) | ≤ 0.2 % |
Ninhydrin-positive subst. (NH4 (570 nm)) | ≤ 0.02 % |
Ninhydrin-positive substances (total) | ≤ 0.5 % |
Cl (Chloride) | ≤ 200 ppm |
SO4 (Sulphate) | ≤ 300 ppm |
Fe (Iron) | ≤ 20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...