Florisil®, feines Pulver <200 mesh, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

288705-250G 288705-1KG
288705-250GEA 110 EUR
288705-250G 288705-1KG
Florisil®, feines Pulver <200 mesh, Supelco®
Magnesiumsilikat
Formel: MgO₃Si
MDL: MFCD00078249
CAS-Nummer: 1343-88-0
EINECS: 215-681-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7185f1253a0288e3ff39ccac6243db3984fe6de6&vid=7ba8b51c22ac5608f10c5b1ec5b0df6942a86b2a&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR