Hexachloroplatinsäure Hexahydrat, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Hydrogenhexachloroplatinat(IV) Hexahydrat, Dihydrogenhexachloroplatinat(IV) Hexahydrat

2890-03EA 346.26 EUR
2890-03
Hexachloroplatinsäure Hexahydrat, BAKER ANALYZED® ACS, J.T.Baker®
Hexachloroplatinsäure Hexahydrat
Formel: H₂PtCl₆·6H₂O
Schmelzpunkt: 150 °C
Dichte: 2,434 g/cm³ (20 °C)
MDL: MFCD00149910
CAS-Nummer: 18497-13-7
EINECS: 241-010-7
UN: 2507
ADR: 8,III
Merck Index: 13,07609

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use Passes Test
Solubility in Alcohol Passes Test
Assay (as Pt) ≥37.50 %
Alkalies and Other Salts (as SO₄) ≤0.05 %
Suitability for Potassium Determination Passes Test
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR