Fraktionssammler, Frac30
Lieferant: Cytiva
29-0230-51
29-0230-51EA 1742 EUR
29-0230-51
Fraktionssammler, Frac30
Fraktionssammler
Frac30 is a round fraction collector designed for use with the ÄKTA start purification system. The Frac30 is controlled via the ÄKTA start touch screen instrument display or using UNICORN start software on a computer. The Frac30 is attached to ÄKTA start for power and communication. It is not suited for standalone operation.
  • Collect up to 30 fractions
  • Support four tube sizes from 1,5 to 15 ml
  • Fractions can be collected in volumes ranging from 0,5 to 15 ml
  • The entire fraction collector rack can be transferred as required