UNICORN™ start, ÄKTA™ start control software, version 1.3
Lieferant: Cytiva
29-7042-34
29-7042-34EA 1444 EUR
29-7042-34
UNICORN™ start, ÄKTA™ start control software, version 1.3
Software Labor-Software
UNICORN™ start is ÄKTA™ start control software. Version 1.3 is the current version. If you have an earlier version, contact us to upgrade.
  • Simple and flexible method creation using predefined templates and phases (steps)
  • Enables real-time monitoring of the run and an intuitive way to control the run
  • Evaluate, compare, and share results. Create and print PDF reports
  • Robust database storage with easy access to data, data security, and data integrity
UNICORN™ start software includes four different modules: System Control, Method Editor, Evaluation, and Administration, allowing you to design runs, operate the ÄKTA™ start instrument, and to evaluate and share results. While ÄKTA™ start can function as a stand-alone system, UNICORN™ start software maximizes the performance of the instrument.