Rotation joint grease, ALSIROL
Lieferant: POLLATH
005/050
291-1320EA 31.2 EUR
291-1320
Rotation joint grease, ALSIROL
Schmiermittel
Conceived as vacuum and high vacuum grease.
  • Scope of application: –40 to 200 °C
  • Resistant to acids and lyes
  • Vapour pressure: 0,00001 mbar
Silicon-free, no silicon derivates. No polymerisation, no sticking. Excellent adhesion and viscosity. Locked joint pairs loosened by short warming. Glass repairs are possible. Not hazardous to health in normal use. Rotation joint grease for sealing stiring systems with glass an metal stirrers. Protects rotating, precision-cut sleeves and stirring shafts from glass abrasion. Also lubricates rotary glass and metal parts.