Wasser, Biotechreagenz zur Injizierung (WFI), J.T.Baker®
Lieferant: Avantor
Filtriert über 0,2 µm-Filter
Avantor’s Water for Injection (WFI) quality water can help supplement existing media and buffer preparation processes by providing a reliable, high-quality alternative to in-house water production. Our water for injection quality water is manufactured under cGMP manufacturing guidelines using a multi-effect distillation process, ozone sparge and UV treatment. It is passed through a 0,2 µm filter prior to filling.
- Manufactured in accordance with ICH-Q7 cGMP guidelines
- Meets USP and EP specifications
- Packaged in bulk (200 L) for commercial use
Bitte beachten For manufacturing, processing, or repackaging. Not intended for parenteral use.
Formel:
H₂O Siedepunkt: 100 °C (1013 hPa) Schmelzpunkt: 0 °C Dichte: 1 g/cm³ (4 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 |
Spezifikation Testergebnisse
Appearance (Clear & Colorless liquid) | Passes Test |
Bacterial Endotoxins (EU/mL) | <=0.25 EU/mL |
Bioburden | <=10 CFU/mL |
pH @ 25° C | 5 to 7 |
Total Organic Carbon (TOC) recorded from recirculation loop | <200 ppm |
Nitrates (NO3) | <0.2 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...