1,1′-Diethyl-4,4′-cyanine iodide, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
1,1′-Diethyl-4,4′-cyanine iodide is a well-known photographic sensitizing dye. It belongs to the class of quinocyanine dyes and exhibits red and blue shifted absorption bands on aggregation. It is also known as 1-ethyl-4-[(1-ethyl-4(1H)-quinolinylidene) methyl] quinolinium iodide. It is suitable for use in silver halide photography.
Formel:
C₂₃H₂₃IN₂ |
MDL:
MFCD00151338 CAS-Nummer: 4727-49-5 |
Spezifikation Testergebnisse
Appearance (Colour) | Green to Very Dark Green |
Appearance (Form): Powder or Crystals | Conforms to Requirements |
Infrared spectrum | Conforms to Structure |
Wavelength, c = 0.003 g/L; Ethanol | 588 - 596 nm |
Extinction Coefficient | ≥75000 |
Carbon | 58.7 - 62.9% |
Nitrogen | 5.9 - 6.4% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...