Kaliumacetat 99,0-100,5% (Trockenmasse), körnig USP, J.T.Baker®

Lieferant: Avantor

Synonyme: Essigsäure Kaliumsalz

2914-01EA 116.93 EUR
2914-01 2914-05 2914-R
Kaliumacetat 99,0-100,5% (Trockenmasse), körnig USP, J.T.Baker®
Kaliumacetat

  • GMP manufactured product

Formel: CH₃COOK
Schmelzpunkt: 304 °C
Dichte: 1,57 g/cm³ (25 °C)
Flammpunkt: >250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012458
CAS-Nummer: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 99.0 - 100.5%
Meets U.S.P Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging ≤1.0%
Bulk Pharmaceutical Chemical 7.5 - 8.5
USP - Assay (CH₃COOK) (dried basis) 99.0 - 100.5 %
USP - Identification Passes Test
USP - Loss on Drying ≤1.0 %
USP - pH (1 in 20) 7.5 - 8.5
USP - Residual Acetic Acid, For Information Only
USP - Sodium (Na) ≤0.03 %
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR