(N-Succinimidyloxycarbonylmethyl)tris(2,4,6-trimethoxyphenyl)phosphonium bromide Acylierungsreagenz, zur Proteinsequenzierung by MALDI-MS, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

29208-50MG
29208-50MGEA 189 EUR
29208-50MG
(N-Succinimidyloxycarbonylmethyl)tris(2,4,6-trimethoxyphenyl)phosphonium bromide Acylierungsreagenz, zur Proteinsequenzierung by MALDI-MS, Supelco®
(N-Succinimidyloxycarbonylmethyl)tris(2,4,6-trimethoxyphenyl)phosphonium bromide

N-Succinimidyloxycarbonylmethyl tris(2,4,6-trimethoxyphenyl)phosphonium bromide (TMPP-Ac-OSu) is an acylating reagent, and is generally used for the selective derivatization of N-terminal a-amino group of peptide.


The TMPP-acetylated peptides undergo fragmentation thereby creating a-type ions, thus obtaining sequence information becomes easier, even without the usage of automated computational approach. It can be used as a labelling reagent in liquid chromatography-coupled mass spectrometry (LC-MS/MS) of N-terminal peptides, so as to improve ionization efficiency, simplify fragmentation, and retention times, thereby leading to better separation of peptides.


TMPP-Ac-OSu was used for labelling peptides, in tandem mass spectrometry, done for converting arginine to ornithine, in order to improve the fragmentation pattern of peptides.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₃₃H₃₉BrNO₁₃P
Molecular Weight: 768,54 g/mol
MDL: MFCD03095505
CAS-Nummer: 226409-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR