Zinkoxid ≥99,0%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Achtung

29211.298EA 133 EUR
29211.298 29211.460
Zinkoxid ≥99,0%, AnalaR NORMAPUR® analytisches Reagens
Zinkoxid
Formel: ZnO
Schmelzpunkt: 1975 °C
Dichte: 5,61 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011300
CAS-Nummer: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.0 %
Insolubility in diluted HCl Max. 50 ppm
Not precipitated by (NH4)2S (as SO4) Max. 0.25 %
Substances reducing KMnO4 (as O) Max. 15 ppm
Cl (Chloride) Max. 10 ppm
NO3 (Nitrate) Max. 20 ppm
PO4 (Phosphate) Max. 20 ppm
SO4 (Sulphate) Max. 100 ppm
As (Arsenic) Max. 1 ppm
Cu (Copper) Max. 20 ppm
Fe (Iron) Max. 5 ppm
Mn (Manganese) Max. 10 ppm
Pb (Lead) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR