2,4-Dimethoxytoluol, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 2,4-Dimethoxy-1-methylbenzene

M-Clarity™ Qualitätsstufe = MQ100

292559-1G 292559-5G
292559-1GEA 104 EUR
292559-1G 292559-5G
2,4-Dimethoxytoluol, Sigma-Aldrich®
2,4-Dimethoxytoluol
Formel: C₉H₁₂O₂
Molecular Weight: 152.1904 g/mol
Siedepunkt: 110...120 °C/30 mmHg (Lit.)
Dichte: 1,036 g/ml (25 °C) (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00010749
CAS-Nummer: 38064-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d135d6cc0e424d4bc897cb6bd923bd70f2ef06e4&vid=d135d6cc0e424d4bc897cb6bd923bd70f2ef06e4&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Colorless
Appearance (Form) Liquid
Infrared spectrum Conforms to Structure
Purity (GC) ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR