Wiederverschliessbares Druckverschlussband SJ387B, Dual Lock™
Lieferant: 3M
SJ387B
293-0009EA 63.9 EUR
293-0009
Wiederverschliessbares Druckverschlussband SJ387B, Dual Lock™
Klebstoffe
This reclosable fastener is the convenient alternative to traditional fastening methods, such as screws, nuts, or bolts. It is designed to deliver a durable, reclosable fastening solution (which can be opened and closed multiple times) that bonds to a variety of substrates including metals, powder-coated paints, and plastics such as acrylics, polycarbonate, and ABS.
  • Snap-in-place installation replaces screws, nails, snaps, and rivets
  • Closes with an audible snap
  • Five times the tensile strength of typical hook-and-loop fasteners
  • Moisture and UV resistant
  • Helps reduce vibration
  • Suitable for both indoor and outdoor use
This can be opened and closed over and over, up to 1000 times before losing 50% of the original tensile strength for repeated access or repositioning. The mushroom heads easily slide over each other allowing positioning of parts before they are snapped together to create a firm fastening attachment. When mated, the fasteners are hidden from view and provide an unbroken surface to help create a smooth, clean appearance. The Type 250 stem density polyolefin fastener mates to Type 170, 250, or 400 giving the user the option of different closure strength depending upon applications.