Autoclaving wraps, Super Polyfoil™, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F132000066
293-2121EA 118 EUR
293-2121
Autoclaving wraps, Super Polyfoil™, SP Bel-Art
Sterilisationsvliese
Polyester backed foil provides a chemically inert barrier to cover vessels for autoclaving, incubating or storage.
  • Thermoplastic layer is laminated
  • Folds easily and flattens without kinking
  • Resistant to punctures, tears and corrosion
  • Polyester film blocks pinholes and resists organic solvents and caustic agents
Standards: Complies with FDA for food contact applications