Wiederlösbare Verschlüsse, SJ352D
Lieferant: 3M
SJ352D
293-2200EA 152 EUR
293-2200
Wiederlösbare Verschlüsse, SJ352D
Klebebänder
Use the 3M™ reclosable fastener SJ352D for a range of applications in a wide variety of industries. The quick fastening feature can help reduce production time and costs. This box offers 5 m of both hook and loop systems, SJ3526N and SJ3527N.
  • Simple attachment alternative to a wide variety of openings and closures, including zips, screws, snaps, hooks, and more
  • Reliable hold for light-duty attachments
  • Opens and closes thousands of times without losing tensile strength
  • Best suited for indoor use
  • Rubber back material
The 3M™ reclosable fastener SJ352D is coated with a high performance rubber adhesive and offers a medium temperature performance up to 50 °C long term (70 °C short term). It's designed for indoor use, bonding well to paper and cardboard, paint, bare and painted metal, glass, sealed wood, and many plastics. Suitable for a range of industries such as transportation, electronics, office equipment, shop fitting, and both internal and external signals.