4-Nitrobenzoldiazoniumtetrafluoroborat 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

294438-10G
294438-10GEA 142 EUR
294438-10G
4-Nitrobenzoldiazoniumtetrafluoroborat 97%, Sigma-Aldrich®
4-Nitrobenzoldiazoniumtetrafluoroborat

4-Nitrobenzenediazonium tetrafluoroborate has been used in ellipsometry. It has also been used for surface modification of carbon nanotubes (CNTs).


4-Nitrobenzenediazonium tetrafluoroborate, also known as Fast Red GG is a Diazonium salt/diazo dye. It is commonly used for the histochemical detection of bilirubin and indolamines.

Formel: C₆H₄BF₄N₃O₂
Molecular Weight: 236,92 g/mol
Schmelzpunkt: 144,..148 °C (Zersetzung)
Dichte: 1,66 g/cm³ (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00012005
CAS-Nummer: 456-27-9
EINECS: 207-261-1
ADR: 8,2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e78f68acb553700ade6aa42f9ea0877398f0a60&vid=a25c018e39edcffca6523f8afcd6abb109d22ac7&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR