2',4'-Dihydroxy-3'-methylacetophenon 90%, technische Qualität, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
1-(2,4-Dihydroxy-3-methylphenyl)ethan-1-on
M-Clarity™ Qualitätsstufe = MQ100
296244-5G
296244-5GEA
118
EUR
296244-5G
2',4'-Dihydroxy-3'-methylacetophenon 90%, technische Qualität, Sigma-Aldrich®
2',4'-Dihydroxy-3'-methylacetophenon
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol Schmelzpunkt: 153…156 °C (Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00010817 CAS-Nummer: 10139-84-1 |
Spezifikation Testergebnisse
Appearance | Yellow to orange to brown powder or crystals |
Infrared spectrum | Conforms to structure |
Gas liquid |
88.0% (minimum) Approved September 12, 2008 RJM |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...