Kaliumiodid AST, 1 g, Tabletten

Lieferant: VWR Chemicals

Synonyme: Iodkalium

296322EEA 83.9 EUR
296322E
Kaliumiodid AST, 1 g, Tabletten
Kaliumiodid
Formel: Kl
Molecular Weight: 166 g/mol
Siedepunkt: 1330
Schmelzpunkt: 681 °C
Dichte: 3.13 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011405
CAS-Nummer: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Bestellen

Spezifikation Testergebnisse

Appearance White biconvex tablets
Weight uniformity: -
Max. 10 % outside 950–1050 mg Conforms
None outside 930–1070 mg Conforms
Average tablet weight 980 --> 1020 mg
Diameter 11.0 --> 11.5 mm
Hardness (kp) (off machine) Min. 8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR