Uniblue A sodium salt ≥70% (durch HPLC) Reactive stain for proteins, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Acid blue 215

298409-1G
298409-1GEA 207 EUR
298409-1G
Uniblue A sodium salt ≥70% (durch HPLC) Reactive stain for proteins, Sigma-Aldrich®
Uniblue A sodium salt
Formel: C₂₂H₁₅N₂NaO₇S₂
Molecular Weight: 506,48 g/mol
MDL: MFCD0001079
CAS-Nummer: 14541-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e8002d1883dfba1236a0d4d1b306848da0966b6&vid=4e8002d1883dfba1236a0d4d1b306848da0966b6&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour): Blue to Very Dark Blue to Very Dark Purple Conforms to Requirements
Appearance (Form) Powder
Infrared Spectrum Conforms to Structure
Wavelength, c = 0.01 g/L; Water 592 - 598 nm
Extinction Coefficient ≥23000
Wavelength 224 - 230 nm
Carbon ≥35.5%
Nitrogen ≥3.5%
Purity (HPLC) ≥70.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR