Eintauchkorb
Lieferant: ENTEGRIS
D10-0315
300-0235EA 0 EUR
300-0235
Eintauchkorb
Probenehmer Probenschöpfer
Dipper basket used to clean or etch small pieces.
  • Hook on end of handle
  • 100 openings of 5,6 mm
  • Fits inside 400 ml and larger beakers
Bitte beachten Articles may not be available in all countries. Please contact your local Avantor office.