1-Butanol 99,4 GC, corrected for water content, AR® ACS, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
|
Achtung
|
Synonyme:
1-Butylalkohol, 1-Hydroxybutan, n-Butanol, Propylcarbinol
3000-26EA
3602.43
EUR
3000-26
1-Butanol 99,4 GC, corrected for water content, AR® ACS, Macron Fine Chemicals™
1-Butanol
Formel:
H₃C(CH₂)₂CH₂OH Molecular Weight: 74.12 g/mol Siedepunkt: 117.6 °C (1013 hPa) Schmelzpunkt: –89 °C Dichte: 0.812 g/cm³ (20 °C) Flammpunkt: 37 °C (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002964 CAS-Nummer: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3 III REACH: 01-2119484630-38 Merck Index: 13,01539 |
|
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | — |
For Laboratory,Research,or Manufacturing Use |
|
ACS - Assay (CH₃CH₂CH₂CH₂OH) (by GC, corrected for water) | ≥99.4 % |
ACS - Butyl Ether (C₈H₁₈O)(by GC) | ≤0.2 % |
ACS - Carbonyl Compounds (as Butyraldehyde) | ≤0.01 % |
ACS - Color (APHA) | ≤10 |
ACS - Residue after Evaporation | ≤0.005 % |
ACS - Titrable Acid (as CH₃COOH) (meq/g) | ≤0.0008 |
ACS - Water (H₂O)(by Karl Fischer titrn) | ≤0.1 % |
Appearance (clear, colorless liquid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...