3,3'-Diaminobenzidin Tetrahydrochlorid (DAB) Kit Peroxidase-Substrat

Lieferant: Biotium
Gefahr

Synonyme: 3,3',4,4'-Biphenyltetramine tetrahydrochloride, DAB HCl

30015
30015.EA 165 EUR
30015.
3,3'-Diaminobenzidin Tetrahydrochlorid (DAB) Kit Peroxidase-Substrat
3,3'-Diaminobenzidin Tetrahydrochlorid

DAB (3,3′-diaminobenzidine) reacts with HRP in the presence of peroxide to yield an insoluble brown-colored product at locations where peroxidase-conjugated antibodies are bound to samples. The brown precipitate is insoluble in alcohol and other organic solvents, making it an excellent substrate for immunohistochemical staining that requires the use of traditional counterstains and mounting media.


The DAB substrate kit includes 20X DAB substrate solution and 20X stabilized peroxide buffer, 15 mL each.


Store at 4 °C. Product is stable for at least 12 months from date of receipt when stored as recommended

Formel: C₁₂H₁₈Cl₄N₄
Molecular Weight: 360,11 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00012969
CAS-Nummer: 7411-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR