Steady-Luc™ Firefly HTS assay kit
Lieferant: Biotium
30028-2 30028-1 30028-3 30028-T 99923
30028-2EA 942 EUR
30028-2 30028-1 30028-3 30028-T 99923.
Steady-Luc™ Firefly HTS assay kit
Tests Zelltests
Firefly luciferase is widely used as a transcriptional reporter gene for research and drug screening. The Steady-Luc™ HTS kit can be used with luminescence plate readers without sample injection systems or for high-throughput screening.