Steady-Luc™ Firefly HTS assay kit
Lieferant: Biotium
30028-2
30028-1
30028-3
30028-T
99923
30028-2EA
942
EUR
30028-2
30028-1
30028-3
30028-T
99923.
Steady-Luc™ Firefly HTS assay kit
Tests
Zelltests
Firefly luciferase is widely used as a transcriptional reporter gene for research and drug screening. The Steady-Luc™ HTS kit can be used with luminescence plate readers without sample injection systems or for high-throughput screening.