Ammoniumhydrogencarbonat 21,30 - 21,73% NH3 basis, powder, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Ammoniumbicarbonat

3003-01EA 0 EUR
3003-01
Ammoniumhydrogencarbonat 21,30 - 21,73% NH3 basis, powder, BAKER ANALYZED®, J.T.Baker®
Ammoniumhydrogencarbonat

Powder.

Formel: NH₄HCO₃
Molecular Weight: 79.06 g/mol
Schmelzpunkt: 35
Dichte: 1.57 g/cm³ (20 °C)
MDL: MFCD00012138
CAS-Nummer: 1066-33-7
EINECS: 213-911-5
Merck Index: 13,00497

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 21.30 - 21.73 %
Assay (as NH₃) 21.30 - 21.73 %
pH of 5% Solution at 25°C 7.0 - 8.0
Nonvolatile Matter ≤0.02 %
Insoluble Matter ≤0.005 %
Nitrate (NO₃) ≤0.002 %
Sulfur Compounds (as SO₄) ≤0.002 %
Chloride (Cl) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR