1,2-Dichloro-4,5-dinitrobenzene ≥98%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
300365-5G
300365-5GEA
286
EUR
300365-5G
1,2-Dichloro-4,5-dinitrobenzene ≥98%, Sigma-Aldrich®
1,2-Dichloro-4,5-dinitrobenzene
1,2-Dichloro-4,5-dinitrobenzene,98%
Formel:
C₆H₂Cl₂N₂O₄ Schmelzpunkt: 105…107 °C (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010865 CAS-Nummer: 6306-39-4 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...