Apoptosis assay kit, NucView® 488 and MitoView™ 633
Lieferant: Biotium
30062
30062.EA 629 EUR
30062.
Apoptosis assay kit, NucView® 488 and MitoView™ 633
Tests Zelltests
NucView® 488 and MitoView™ 633 kit allows profiling of apoptotic cells simultaneously with changes in mitochondrial membrane potential using either fluorescence microscopy or flow cytometry.
The bi-functional NucView® 488 caspase-3 substrate can detect both intracellular caspase-3 and at the same time stain the cell nucleus, which is known to undergo morphological changes during apoptosis. MitoView™ 633 is a far-red fluorescent mitochondrial dye. The staining is dependent upon the mitochondrial membrane potential; thus, apoptotic cells exhibit a much lower MitoView™633 dye staining compared to healthy cells. Furthermore, the spectral separation of these two dyes minimises fluorescence overlap.
The optimal detection settings for MitoView™ 633 are the same as for Cy®5 and other far-red dyes. However, the dye also has visible red fluorescence and can be imaged using Cy®3 settings as well. Consequently, the dye cannot be used for two-colour imaging with other red probes.
Lieferumfang: Kit contains NucView®488 caspase-3 substrate and the MitoView™ 633 mitochondrial dye sufficient for 100 assays.