Flußsäure ≥48%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™

Lieferant: Honeywell Chemicals
Gefahr

30107-1L 30107-500ML 30107-5L 30107-6X1L
30107-1LEA 85.39 EUR
30107-1L 30107-500ML 30107-5L 30107-6X1L
Flußsäure ≥48%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Hydrofluoric acid
Formel: HF
Siedepunkt: 105 °C (1,013 hPa)
Schmelzpunkt: –35 °C
Dichte: 1,170 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011346
CAS-Nummer: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Assay Min. 48 %
Residue on ignition (SO₄) Max. 0.0005 %
Silver (Ag) Max. 0.000002 %
Aluminium (Al) Max. 0.000005 %
Arsenic (As) Max. 0.000005 %
Barium (Ba) Max. 0.00001 %
Beryllium (Be) Max. 0.000002 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000001 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00002 %
Germanium (Ge) Max. 0.000005 %
Potassium (K) Max. 0.00001 %
Lithium (Li) Max. 0.000002 %
Magnesium (Mg) Max. 0.00002 %
Manganese (Mn) Max. 0.000005 %
Molybdenum (Mo) Max. 0.000005 %
Sodium (Na) Max. 0.00002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000005 %
Strontium (Sr) Max. 0.000002 %
Titanium (Ti) Max. 0.00001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000005 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Heavy metals (as Pb) Max. 0.00005 %
Chloride (Cl) Max. 0.0001 %
Hexafluorosilicic acid (H₂SiF₆) Max. 0.005 %
Phosphate (PO₄) Max. 0.00005 %
Sulfate (SO₄) Max. 0.0002 %
Sulfite (SO₃) Max. 0.0002 %
Vapor Density 1.27 (vs air)
Appearance (color) complying
Appearance (form) complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR