1,3-Propandiylbis(diphenylphosphin) -dichlornickel

Lieferant: Thermo Fisher Scientific
Achtung

30167.09
30167.09EA 166 EUR
30167.09
1,3-Propandiylbis(diphenylphosphin) -dichlornickel
1,3-Bis(diphenylphosphino)propanenickel(II) chloride

CAS No.: 15629-92-2 Molecular Formula: C27H26Cl2NiP2 Formula Weight: 542.03 Physical Form: Powder Melting Point: 213° dec. MDL No.: MFCD00015318

Formel: C₂₇H₂₆Cl₂NiP₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00015318
CAS-Nummer: 15629-92-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR