Dimethyltitanocene 5% (w/w) in Toluol, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

Synonyme: Petasis Reagent, Bis(cyclopentadienyl)dimethyltitanium

301761000
301761000.EA 388 EUR
301761000.
Dimethyltitanocene 5% (w/w) in Toluol, AcroSeal®
Dimethyltitanocene

Appearance: Orange to dark red Solution

Formel: C₁₂H₁₆Ti
Molecular Weight: 208,13 g/mol
Dichte: 0,9 g/cm³
Lagertemperatur: Gefrierschrank
MDL: MFCD02093882
CAS-Nummer: 1271-66-5
UN: 1294
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Appearance Orange to dark red Solution
NMR (1H-NMR) Consistent with structure
Strength 5 % (approximately)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR