Kaliumchlorat ≥99,0% (durch KMnO₄-Titration), Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
3024-07EA
0
EUR
3024-07
Kaliumchlorat ≥99,0% (durch KMnO₄-Titration), Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Potassium chlorate
Formel:
KClO₃ Molecular Weight: 122.55 g/mol Siedepunkt: 400 °C (1013 hPa) Schmelzpunkt: 356 °C Dichte: 2.34 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011361 CAS-Nummer: 3811-04-9 EINECS: 223-289-7 UN: 1485 ADR: 5.1 II Merck Index: 13,07703 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (KClO₃) (by KMnO₄ titrn) | ≥99.0 % |
Insoluble Matter | ≤0.005 % |
pH of 5% Solution at 25°C, For Information Only | For Information Only |
Bromate (BrO₃) | ≤0.01 % |
Calcium (Ca) | ≤0.002 % |
Chloride (Cl) | ≤0.001 % |
ACS - Magnesium (Mg) | ≤0.002 % |
Sulfate (SO₄) | Passes Test |
Sodium (Na) | ≤0.010 % |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤2.0 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...