D(-)-Sorbit 97,0-100,5, GPR RECTAPUR®

Lieferant: VWR Chemicals

Synonyme: D(-)-Glucit, (2R,3R,4R,5S)-(-)-1,2,3,4,5,6-Hexanhexol, D(-)-Sorbitol, D(-)-Glucitol

302424AEA 124 EUR
302424A 302425B 302426C
D(-)-Sorbit 97,0-100,5, GPR RECTAPUR®
D(-)-Sorbit
Formel: HOH₂C(CH(OH))₄CH₂OH
Molecular Weight: 182.17 g/mol
Siedepunkt: 295
Schmelzpunkt: 94…98 °C
Dichte: 1.524 g/cm³ (20 °C)
Flammpunkt: >100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004708
CAS-Nummer: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (calculated on anhydrous) 97.0- 100.5 %
Appearance of solution (10 %; water) Passes test
Identification Passes test
Spec. opt. rot.(10 %; borate; anhydrous) 4.0 - 7.0 °
Reducing sugars (calculated as C6H12O6) Max. 0.2 %
Ignition residue (SO4) Max. 0.1 %
Water Max. 1.5 %
Cl (Chloride) Max. 50 ppm
SO4 (Sulphate) Max. 100 ppm
Ni (Nickel) Max. 1 ppm
Pb (Lead) Max. 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR