Ammoniumthiocyanat 0.05 mol/l (0.05 N) in wässriger Lösung
Lieferant: VWR Chemicals
Synonyme:
Ammoniumsulfocyanid, Ammoniumrhodanid
302635.1000EA
35
EUR
302635.1000
Ammoniumthiocyanat 0.05 mol/l (0.05 N) in wässriger Lösung
Ammoniumthiocyanat
Formel:
NH₄SCN Molecular Weight: 76,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011428 CAS-Nummer: 1762-95-4 EINECS: 217-175-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...