Trypanblau 60% (Farbstoffgehalt) ≥80% (durch HPLC), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)

M-Clarity™ Qualitätsstufe = MQ200

302643-100G 302643-25G
302643-100GEA 260 EUR
302643-100G 302643-25G
Trypanblau 60% (Farbstoffgehalt) ≥80% (durch HPLC), Sigma-Aldrich®
Trypanblau

Trypan Blue (C.l. 23850) is a large, tetrasulfonated anionic, hydrophilic dye. It is also known as diamine blue 3B. Trypan blue has been used as a dye in trypan blue exclusion assay/cell viability assay to detect dead cells.

Formel: C₃₄H₂₄N₆Na₄O₁₄S₄
Molecular Weight: 960.82 g/mol
Schmelzpunkt: >300 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00003969
CAS-Nummer: 72-57-1
EINECS: 200-786-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6325edea40d1e0e997dcdcecb46a3e7f1cb92726&vid=6325edea40d1e0e997dcdcecb46a3e7f1cb92726&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Dark Green to Brown or Brown-Black
Appearance (Form) Powder or Crystals or Granules
Infrared spectrum Conforms to Structure
Wavelength 603 - 607 nm
Extinction Coefficient, c = 0.01 g/L; Methanol ≥60000
Carbon ≥24.6 %
Nitrogen ≥5.0 %
Purity (HPLC) ≥80.0 %
Solubility (Turbidity); Opaque, c = 1mg/ml; Water Conforms
Solubility (Color) Dark Blue
Recommended Retest Period 5 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR