Reaktionsgefäße, Superslik™
Lieferant: LABCON
3039-561-000
3039-561-000EA 0 EUR
3039-561-000
Reaktionsgefäße, Superslik™
Röhrchen Reaktionsgefäße
PP, translucent. For use with rDNA and viscous proteins, these nonstick, graduated, polypropylene tubes may be used between –90 and +140 °C.
  • Leakproof snap caps
  • Non sterile
  • Nonsticking
  • Graduated markings
They feature flat caps and frosted areas on the sides for easy labelling. Leakproof snap caps may be penetrated by a syringe. Easy-to-open caps will not pop open during autoclaving. The 0,5 ml tubes fit many thermal cyclers. All tubes are non sterile. RNase- and DNase-free.