Trimethyl thiophosphate

Lieferant: Thermo Fisher Scientific
Gefahr

30395.03 30395.06 30395.14
30395.03EA 31.4 EUR
30395.03 30395.06 30395.14
Trimethyl thiophosphate
Trimethyl thiophosphate

CAS No.: 152-18-1 Molecular Formula: (CH3O)3P(S) Formula Weight: 156.15 Physical Form: Liquid Appearance: Off-white Odour: Sulphurous UN No.: UN3278 MDL No.: MFCD00014888

Formel: C₃H₉O₃PS
Lagertemperatur: Raumtemperatur
MDL: MFCD00014888
CAS-Nummer: 152-18-1
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR