Trimethyl thiophosphate
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
30395.03
30395.06
30395.14
30395.03EA
31.4
EUR
30395.03
30395.06
30395.14
Trimethyl thiophosphate
Trimethyl thiophosphate
CAS No.: 152-18-1
Molecular Formula: (CH3O)3P(S)
Formula Weight: 156.15
Physical Form: Liquid
Appearance: Off-white
Odour: Sulphurous
UN No.: UN3278
MDL No.: MFCD00014888
Formel:
C₃H₉O₃PS Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014888 CAS-Nummer: 152-18-1 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...