Kaliumchlorid 99,0-100,5% (durch argentometrische Titration), Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
Achtung
|
Synonyme:
Potassiumchloride
3040-09EA
1738.1
EUR
3040-09
3040-05
3040-01
3040-07
3040-R
Kaliumchlorid 99,0-100,5% (durch argentometrische Titration), Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Potassium chloride
Formel:
KCl Molecular Weight: 74.55 g/mol Siedepunkt: 1420 °C (1013 hPa) Schmelzpunkt: 773 °C Dichte: 1.99 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011360 CAS-Nummer: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,07621 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | 99.0 - 100.5% |
For Laboratory,Research,or Manufacturing Use | ≤1.0% |
Assay (KCl) (by Ag titrn) | 99.0 - 100.5 % |
Loss on Drying at 105°C | ≤1.0 % |
Insoluble Matter | ≤0.005 % |
pH of 5% Solution at 25°C | 5.4 - 8.6 |
Iodide (I) | ≤0.002 % |
Bromide (Br) | ≤0.01 % |
Calcium (Ca) | ≤0.002 % |
Chlorate and Nitrate (as NO₃) | ≤0.003 % |
Magnesium (Mg) | ≤0.001 % |
Nitrogen Compounds (as N) | ≤0.001 % |
Sulfate (SO₄) | ≤0.001 % |
Barium (Ba) | Passes Test |
Sodium (Na)(by FES) | ≤0.005 % |
Phosphate (PO₄) | ≤5 ppm |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Arsenic (As) | ≤1 ppm |
Trace Impurities - Iron (Fe) | ≤2.0 ppm |
Trace Impurities - Nickel (Ni) | ≤1.000 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...