Kaliumchlorid 99,0-100,5% (durch argentometrische Titration), Kristalle, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Potassiumchloride

3040-09EA 1738.1 EUR
3040-09 3040-05 3040-01 3040-07 3040-R
Kaliumchlorid 99,0-100,5% (durch argentometrische Titration), Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Potassium chloride
Formel: KCl
Molecular Weight: 74.55 g/mol
Siedepunkt: 1420 °C (1013 hPa)
Schmelzpunkt: 773 °C
Dichte: 1.99 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011360
CAS-Nummer: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 99.0 - 100.5%
For Laboratory,Research,or Manufacturing Use ≤1.0%
Assay (KCl) (by Ag titrn) 99.0 - 100.5 %
Loss on Drying at 105°C ≤1.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 5.4 - 8.6
Iodide (I) ≤0.002 %
Bromide (Br) ≤0.01 %
Calcium (Ca) ≤0.002 %
Chlorate and Nitrate (as NO₃) ≤0.003 %
Magnesium (Mg) ≤0.001 %
Nitrogen Compounds (as N) ≤0.001 %
Sulfate (SO₄) ≤0.001 %
Barium (Ba) Passes Test
Sodium (Na)(by FES) ≤0.005 %
Phosphate (PO₄) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Arsenic (As) ≤1 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Trace Impurities - Nickel (Ni) ≤1.000 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR