Triethyl-2-phosphonopentanoat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

30413.04 30413.09
30413.04EA 124 EUR
30413.04 30413.09
Triethyl-2-phosphonopentanoat ≥98%
Triethyl-2-phosphonopentanoat

Purity: 98% CAS No.: 35051-49-1 Molecular Formula: C11H23O5P Formula Weight: 266.27 Physical Form: Liquid Appearance: Colourless MDL No.: MFCD00015163

Formel: C₁₁H₂₃O₅P
Siedepunkt: 98 °C (2 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00015163
CAS-Nummer: 35051-49-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR