2-Propanol 99,8, ChromAR® HPLC, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Dimethylcarbinol, IPA, Isopropylalkohol, 2-Hydroxypropan, Isopropanol

3043-23EA 0 EUR
3043-23
2-Propanol 99,8, ChromAR® HPLC, Macron Fine Chemicals™
2-Propanol

Suitable for Liquid Chromatography and UV-Spectrophotometry

Formel: (CH₃)₂CHOH
Molecular Weight: 60.1 g/mol
Siedepunkt: 82 °C (1013 hPa)
Schmelzpunkt: –89 °C
Dichte: 0.786 g/cm³ (20 °C)
Flammpunkt: 12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011674
CAS-Nummer: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (by GC) min. 99.8 %
Assay (GC) min. 99.8 %
Residue after Evaporation max. 0.0005 %
Residue on Evaporation max. 0.0005 %
Titrable Acid (meq/g) max. 0.0005
Titrable Base (meq/g) max. 0.0005
Water (KF) max. 0.05 %
Ultraviolet Absorbance (1.00-cm cell vs water):
at 210 nm max. 0.4
at 220 nm max. 0.2
at 230 nm max. 0.1
at 240 nm max. 0.05
at 250 nm max. 0.02
UV Cut-off max. 203 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR