Strontiumnitrat, wasserfrei ≥99,97% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
30486.22
30486.22EA
365
EUR
30486.22
Strontiumnitrat, wasserfrei ≥99,97% (Metall-Basis)
Strontium nitrate
Purity: 99.97%
CAS No.: 10042-76-9
Molecular Formula: N2O6Sr
Formula Weight: 211.63
Physical Form: Crystalline
Density: 2.990
Appearance: White
Odour: Odourless
Melting Point: 570°
Merck Reference: 14,8845
UN No.: 1507
MDL No.: MFCD00011248
Formel:
Sr(NO₃)₂ Molecular Weight: 211,63 g/mol Siedepunkt: 645 °C (1013 hPa) Schmelzpunkt: 570 °C Dichte: 2,990 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011248 CAS-Nummer: 10042-76-9 EINECS: 233-131-9 UN: 1507 ADR: 5.1,III Merck Index: 14,8845 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...