Iron(II) oxide ≥99,5% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Purity: 99.5%
CAS No.: 1345-25-1
Molecular Formula: FeO (may contain trace amounts of other phases)
Formula Weight: 71.85
Physical Form: -10 Mesh
Density: 5.7
Sensitivity: Air Sensitive
Appearance: Black
Odour: Odourless
Melting Point: 1369°
Solubility: Insoluble in water and alkalies. Soluble in HCl, H2SO4. Slightly soluble in HNO3
MDL No.: MFCD00016095
Applications: In manufacture of green, heat-absorbing glass; in steel manufacture; and as a catalyst
Formel:
FeO Schmelzpunkt: 1369 °C Dichte: 5,7 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016095 CAS-Nummer: 1345-25-1 EINECS: 215-721-8 Merck Index: 14,4052 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...