Iron(II) ethylenediammonium sulphate tetrahydrate ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ferrous Ethylenediamine Sulphate
30547.14
30547.22
30547.14EA
66.2
EUR
30547.14
30547.22
Iron(II) ethylenediammonium sulphate tetrahydrate ≥99%
Iron(II) ethylenediammonium sulphate tetrahydrate
Iron(II) ethylenediammonium sulfate tetrahydrate is used in the preparation of gold nanoparticles by reacting with gold(III)-bis[ethylene-diamine]-chloride.
Purity: 99%
CAS No.: 34962-29-3
Molecular Formula: C2H10FeN2O8S2.4H2O
Formula Weight: 382.14 (310.08anhy)
Physical Form: Crystalline
Appearance: White to pale green
MDL No.: MFCD00082474
Formel:
C₂H₈N₂ Molecular Weight: 310.07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082474 CAS-Nummer: 113193-60-5 EINECS: 264-357-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...