1,10-Phenanthrolineiron(II) perchlorate

Lieferant: Thermo Scientific
Gefahr

30557.09
30557.09EA 255 EUR
30557.09
1,10-Phenanthrolineiron(II) perchlorate
1,10-Phenanthrolineiron(II) perchlorate

CAS No.: 14586-54-0 Molecular Formula: C36H24Cl2FeN6O8 Formula Weight: 795.37 Physical Form: Crystalline Appearance: Red UN No.: UN1479 MDL No.: MFCD00050959


Applications: Oxidation, reduction indicator

Formel: C₃₆H₂₄Cl₂FeN₆O₈
Molecular Weight: 795,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00050959
CAS-Nummer: 14586-54-0
EINECS: 238-627-9
ADR: 5.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR