Bis(1-butylpentyl)decane-1,10-diyl diglutarate ≥97%, Selectophore™ Ionophor, Supelco®

Lieferant: Merck

Synonyme: ETH 469, 5,30-Dibutyl-6,12,23,29-tetraoxatetratriacontane-7,11,24,28-tetrane

M-Clarity™ Qualitätsstufe = MQ100

30585-1G
30585-1GEA 463 EUR
30585-1G
Bis(1-butylpentyl)decane-1,10-diyl diglutarate ≥97%, Selectophore™ Ionophor, Supelco®
Bis(1-butylpentyl)decane-1,10-diyl diglutarate

Bis(1-butylpentyl)decane-1,10-diyl diglutarate, was used for the preparation of optode membranes, which are sodium selective and are based on neutral ionophore, primarily used for the characterization of total sodium concentration in human blood plasma.


It was also used for the preparation of nitrite-selective membrane electrode, used for the potentiometric determination of NAD(P)H coenzymes.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₃₈H₇₀O₈
Molecular Weight: 654.96 g/mol
MDL: MFCD00043109
CAS-Nummer: 101342-76-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1d39fa49e1e5814be4af2d9001481428fd16982&vid=8530a5f049776c45d8593a954f2deabac5444ee2&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR